3-Methoxy-3'-methylbenzhydrol ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 3-Methoxy-α-(3-methylphenyl)benzenemethanol

246841-1G 246841-500MG 246841-5G 246841-25G
77467-410EA 362.56 USD
77467-410 77480-130 77490-440 77500-596
3-Methoxy-3'-methylbenzhydrol ≥95%
3-Methoxy-3'-methylbenzhydrol

Precaución: For research use only

Formula: C₁₅H₁₆O₂
MW: 228.29 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06201276
CAS Number: 844683-32-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR