4-tert-Butyl-3'-methylbenzhydrol ≥95%

Supplier: AOBChem USA

Warning

Synonyms: α-[4-(1,1-Dimethylethyl)phenyl]-3-methylbenzenemethanol

246829-1G 246829-500MG 246829-5G
77467-406EA 648.9 USD
77467-406 77480-126 77490-436
4-tert-Butyl-3'-methylbenzhydrol ≥95%
4-tert-Butyl-3'-methylbenzhydrol

Precaución: For research use only

Formula: C₁₈H₂₂O
MW: 254.37 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06201263
CAS Number: 842140-68-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR