1-Chloro-2-(4-ethoxybenzyl)benzene ≥96%

Supplier: AOBChem USA

Warning

246674-1G 246674-500MG 246674-5G 246674-25G
77467-398EA 231.75 USD
77467-398 77480-118 77490-428 77500-586
1-Chloro-2-(4-ethoxybenzyl)benzene ≥96%
1-Chloro-2-(4-ethoxybenzyl)benzene

Precaución: For research use only

Formula: C₁₅H₁₅ClO
Storage Temperature: Ambient
MDL Number: MFCD28359478
CAS Number: 1662702-90-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR