2,4-Dimethoxybenzophenone ≥95%

Supplier: AOBChem USA

Warning

246600-1G 246600-500MG 246600-5G 246600-25G
77467-394EA 165.83 USD
77467-394 77480-114 77490-424 77500-582
2,4-Dimethoxybenzophenone ≥95%
2,4-Dimethoxybenzophenone

Precaución: For research use only

Formula: C₁₅H₁₄O₃
MW: 242.27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01098030
CAS Number: 3555-84-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR