Cyclobutyl(2,3-dimethylphenyl)methanone ≥95%

Supplier: AOBChem USA

Warning

246511-1G 246511-500MG 246511-5G 246511-25G
77467-390EA 423.33 USD
77467-390 77480-110 77490-420 77500-580
Cyclobutyl(2,3-dimethylphenyl)methanone ≥95%
Cyclobutyl(2,3-dimethylphenyl)methanone

Precaución: For research use only

Formula: C₁₃H₁₆O
Storage Temperature: Ambient
MDL Number: MFCD03841264
CAS Number: 898790-66-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR