1-Bromo-4-cyclopropyl-2-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

24646-1G 24646-500MG 24646-5G 24646-25G
77467-382EA 282.22 USD
77467-382 77480-102 77490-412 77500-572
1-Bromo-4-cyclopropyl-2-methylbenzene ≥97%
1-Bromo-4-cyclopropyl-2-methylbenzene

Precaución: For research use only

Formula: C₁₀H₁₁Br
MW: 211.098 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21099972
CAS Number: 1353854-72-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR