1-(2-Ethylphenyl)propan-1-one ≥96%

Supplier: AOBChem USA

Warning

Synonyms: 1-(2-Ethylphenyl)-1-propanone, 2'-Ethylpropiophenone

246434-1G 246434-500MG 246434-5G 246434-25G
77467-378EA 292.52 USD
77467-378 77480-098 77490-408 77500-568
1-(2-Ethylphenyl)propan-1-one ≥96%
1-(2-Ethylphenyl)propan-1-one

Precaución: For research use only

Formula: C₁₁H₁₄O
MW: 162.23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18396795
CAS Number: 16819-79-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR