α-Cyclopropyl-2-fluoro-α-methylbenzenemethanol ≥97%

Supplier: AOBChem USA

Warning

246186-1G 246186-500MG 246186-5G 246186-25G
77467-374EA 226.6 USD
77467-374 77480-094 77490-404 77500-562
α-Cyclopropyl-2-fluoro-α-methylbenzenemethanol ≥97%
α-Cyclopropyl-2-fluoro-α-methylbenzenemethanol

Precaución: For research use only

Formula: C₁₁H₁₃FO
Storage Temperature: Ambient
MDL Number: MFCD16779262
CAS Number: 1251249-36-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR