α-Cyclopropyl-3-fluoro-α-methylbenzenemethanol ≥97%

Supplier: AOBChem USA

Warning

246117-1G 246117-500MG 246117-5G 246117-25G
77467-366EA 339.9 USD
77467-366 77480-086 77490-396 77500-554
α-Cyclopropyl-3-fluoro-α-methylbenzenemethanol ≥97%
α-Cyclopropyl-3-fluoro-α-methylbenzenemethanol

Precaución: For research use only

Formula: C₁₁H₁₃FO
Storage Temperature: Ambient
MDL Number: MFCD12152954
CAS Number: 1178016-13-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR