1-Cyclopropyl-1-phenylethanol ≥95%

Supplier: AOBChem USA

Warning

Synonyms: α-Cyclopropyl-α-methylbenzenemethanol

246093-1G 246093-500MG 246093-5G 246093-25G
77467-362EA 85.49 USD
77467-362 77480-082 77490-392 77500-550
1-Cyclopropyl-1-phenylethanol ≥95%
1-Cyclopropyl-1-phenylethanol

Precaución: For research use only

Formula: C₁₁H₁₄O
Storage Temperature: Ambient
MDL Number: MFCD00019243
CAS Number: 5558-04-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR