3-Fluoro-5-(methylthio)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

24558-1G 24558-500MG 24558-5G 24558-25G
77467-358EA 227.63 USD
77467-358 77480-078 77490-388 77500-544
3-Fluoro-5-(methylthio)benzaldehyde ≥97%
3-Fluoro-5-(methylthio)benzaldehyde

Precaución: For research use only

Formula: C₈H₇FOS
MW: 170.204 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28142582
CAS Number: 1289051-26-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR