1-Bromo-2-ethoxy-4-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

24523-1G 24523-500MG 24523-5G 24523-25G
77467-354EA 109.18 USD
77467-354 77480-074 77490-384 77500-540
1-Bromo-2-ethoxy-4-methylbenzene ≥97%
1-Bromo-2-ethoxy-4-methylbenzene

Precaución: For research use only

Formula: C₉H₁₁BrO
MW: 215.087 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22543717
CAS Number: 1394291-57-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR