2-Fluoro-3-(methylsulfanyl)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

24488-1G 24488-500MG 24488-5G 24488-25G
77467-346EA 158.62 USD
77467-346 77480-066 77490-376 77500-534
2-Fluoro-3-(methylsulfanyl)phenylboronic acid ≥97%
2-Fluoro-3-(methylsulfanyl)phenylboronic acid

Precaución: For research use only

Formula: C₇H₈BFO₂S
MW: 186.012 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22628197
CAS Number: 1451392-57-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR