3-Bromo-2-fluoro-4-pyridinol ≥96%

Supplier: AOBChem USA

Warning

244365-1G 244365-500MG 244365-5G 244365-25G
77467-338EA 328.57 USD
77467-338 77480-060 77490-368 77500-526
3-Bromo-2-fluoro-4-pyridinol ≥96%
3-Bromo-2-fluoro-4-pyridinol

Precaución: For research use only

Formula: C₅H₃BrFNO
Storage Temperature: Ambient
MDL Number: MFCD28739486
CAS Number: 1807230-95-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR