1-Bromo-2-methoxy-3,5-bis(trifluoromethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

24409-1G 24409-500MG 24409-5G
77467-334EA 851.81 USD
77467-334 77480-056 77490-364
1-Bromo-2-methoxy-3,5-bis(trifluoromethyl)benzene ≥97%
1-Bromo-2-methoxy-3,5-bis(trifluoromethyl)benzene

Precaución: For research use only

Formula: C₉H₅BrF₆O
MW: 323.03 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28727807
CAS Number: 197015-76-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR