3-Chloro-4-(methylthio)phenol ≥97%

Supplier: AOBChem USA

Warning

24357-1G 24357-500MG 24357-5G 24357-25G
77467-330EA 253.38 USD
77467-330 77480-052 77490-360 77500-520
3-Chloro-4-(methylthio)phenol ≥97%
3-Chloro-4-(methylthio)phenol

Precaución: For research use only

Formula: C₇H₇ClOS
MW: 174.648 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16999022
CAS Number: 13560-43-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR