1,2,3-Trifluoro-4-(trimethylsilyl)benzene ≥95%

Supplier: AOBChem USA

Warning

24341-1G 24341-500MG 24341-5G 24341-25G
77467-322EA 168.92 USD
77467-322 77480-044 77490-352 77500-512
1,2,3-Trifluoro-4-(trimethylsilyl)benzene ≥95%
1,2,3-Trifluoro-4-(trimethylsilyl)benzene

Precaución: For research use only

Formula: C₉H₁₁F₃Si
Storage Temperature: Ambient
MDL Number: MFCD27965453
CAS Number: 363598-55-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR