2,4-Dichloro-1-(phenylmethoxy)benzene ≥97%

Supplier: AOBChem USA

Warning

24326-1G 24326-500MG 24326-5G 24326-25G
77467-318EA 256.47 USD
77467-318 77480-042 77490-350 77500-508
2,4-Dichloro-1-(phenylmethoxy)benzene ≥97%
2,4-Dichloro-1-(phenylmethoxy)benzene

Precaución: For research use only

Formula: C₁₃H₁₀Cl₂O
MW: 253.124 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00658357
CAS Number: 19578-69-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR