3-Hydroxy-3-phenylpropionitrile ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-Hydroxy-3-phenylpropanenitrile

242965-1G 242965-5G 242965-25G
77467-306EA 67.98 USD
77467-306 77490-338 77500-506
3-Hydroxy-3-phenylpropionitrile ≥97%
3-Hydroxy-3-phenylpropionitrile

Precaución: For research use only

Formula: C₉H₉NO
MW: 147.174 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00544189
CAS Number: 17190-29-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR