1-(5-Bromo-6-methylpyridin-2-yl)ethanone ≥95%

Supplier: AOBChem USA

Warning

242529-1G 242529-500MG 242529-5G
77467-302EA 457.32 USD
77467-302 77480-026 77490-334
1-(5-Bromo-6-methylpyridin-2-yl)ethanone ≥95%
1-(5-Bromo-6-methylpyridin-2-yl)ethanone

Precaución: For research use only

Formula: C₈H₈BrNO
Storage Temperature: Ambient
MDL Number: MFCD22553309
CAS Number: 1822993-80-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR