2,5-Difluoro-α-heptylbenzenemethanol ≥97%

Supplier: AOBChem USA

Warning

242325-1G 242325-500MG 242325-5G 242325-25G
77467-298EA 415.09 USD
77467-298 77480-022 77490-330 77500-502
2,5-Difluoro-α-heptylbenzenemethanol ≥97%
2,5-Difluoro-α-heptylbenzenemethanol

Precaución: For research use only

Formula: C₁₄H₂₀F₂O
MW: 242.31 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16357519
CAS Number: 1281634-97-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR