2-Chloro-6-fluoro-α-heptylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

242252-1G 242252-500MG 242252-5G 242252-25G
77467-294EA 415.09 USD
77467-294 77480-018 77490-326 77500-498
2-Chloro-6-fluoro-α-heptylbenzenemethanol ≥95%
2-Chloro-6-fluoro-α-heptylbenzenemethanol

Precaución: For research use only

Formula: C₁₄H₂₀ClFO
MW: 258.76 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20967449
CAS Number: 1498245-92-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR