2,6-Difluoro-α-heptyl-3-methylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

242225-1G 242225-500MG 242225-5G 242225-25G
77467-290EA 415.09 USD
77467-290 77480-014 77490-320 77500-494
2,6-Difluoro-α-heptyl-3-methylbenzenemethanol ≥95%
2,6-Difluoro-α-heptyl-3-methylbenzenemethanol

Precaución: For research use only

Formula: C₁₅H₂₂F₂O
Storage Temperature: Ambient
MDL Number: MFCD25807697
CAS Number: 1551030-44-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR