2-Bromo-6-fluoro-α-heptylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

242181-1G 242181-500MG 242181-5G 242181-25G
77467-286EA 415.09 USD
77467-286 77480-010 77490-318 77500-490
2-Bromo-6-fluoro-α-heptylbenzenemethanol ≥95%
2-Bromo-6-fluoro-α-heptylbenzenemethanol

Precaución: For research use only

Formula: C₁₄H₂₀BrFO
MW: 303.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34870665
CAS Number: 1778756-58-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR