3-Bromo-2,6-difluoro-α-heptylbenzenemethanol ≥97%

Supplier: AOBChem USA

Warning

242159-1G 242159-500MG 242159-5G 242159-25G
77467-282EA 415.09 USD
77467-282 77480-006 77490-314 77500-486
3-Bromo-2,6-difluoro-α-heptylbenzenemethanol ≥97%
3-Bromo-2,6-difluoro-α-heptylbenzenemethanol

Precaución: For research use only

Formula: C₁₄H₁₉BrF₂O
Storage Temperature: Ambient
MDL Number: MFCD34870646
CAS Number: 1982087-46-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR