4-Bromo-α-heptylbenzenemethanol ≥97%

Supplier: AOBChem USA

Warning

242111-1G 242111-500MG 242111-5G 242111-25G
77467-278EA 226.6 USD
77467-278 77480-002 77490-310 77500-482
4-Bromo-α-heptylbenzenemethanol ≥97%
4-Bromo-α-heptylbenzenemethanol

Precaución: For research use only

Formula: C₁₄H₂₁BrO
Storage Temperature: Ambient
MDL Number: MFCD18352297
CAS Number: 76287-55-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR