2-Chloro-6-fluoro-α-pentylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

242039-1G 242039-500MG 242039-5G 242039-25G
77467-274EA 397.58 USD
77467-274 77479-998 77490-306 77500-478
2-Chloro-6-fluoro-α-pentylbenzenemethanol ≥95%
2-Chloro-6-fluoro-α-pentylbenzenemethanol

Precaución: For research use only

Formula: C₁₂H₁₆ClFO
MW: 230.71 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21171301
CAS Number: 1484371-28-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR