2-Bromo-6-fluoro-α-pentylbenzenemethanol ≥96%

Supplier: AOBChem USA

Warning

241976-1G 241976-500MG 241976-5G 241976-25G
77467-270EA 284.28 USD
77467-270 77479-994 77490-302 77500-474
2-Bromo-6-fluoro-α-pentylbenzenemethanol ≥96%
2-Bromo-6-fluoro-α-pentylbenzenemethanol

Precaución: For research use only

Formula: C₁₂H₁₆BrFO
MW: 275.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34870593
CAS Number: 1603044-83-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR