5-Bromo-2-fluoro-α-pentylbenzenemethanol ≥97%

Supplier: AOBChem USA

Warning

241930-1G 241930-500MG 241930-5G 241930-25G
77467-266EA 226.6 USD
77467-266 77479-990 77490-298 77500-470
5-Bromo-2-fluoro-α-pentylbenzenemethanol ≥97%
5-Bromo-2-fluoro-α-pentylbenzenemethanol

Precaución: For research use only

Formula: C₁₂H₁₆BrFO
Storage Temperature: Ambient
MDL Number: MFCD21171367
CAS Number: 1462883-89-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR