2,5-Dimethyl-α-pentylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

241910-1G 241910-500MG 241910-5G 241910-25G
77467-262EA 397.58 USD
77467-262 77479-986 77490-294 77500-468
2,5-Dimethyl-α-pentylbenzenemethanol ≥95%
2,5-Dimethyl-α-pentylbenzenemethanol

Precaución: For research use only

Formula: C₁₄H₂₂O
MW: 206.32 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12787275
CAS Number: 854179-27-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR