3-(Methylthio)-α-pentylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

241869-1G 241869-500MG 241869-5G
77467-258EA 597.4 USD
77467-258 77479-982 77490-290
3-(Methylthio)-α-pentylbenzenemethanol ≥95%
3-(Methylthio)-α-pentylbenzenemethanol

Precaución: For research use only

Formula: C₁₃H₂₀OS
Storage Temperature: Ambient
MDL Number: MFCD34870539
CAS Number: 2159917-30-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR