α-Hexyl-2,4-dimethylbenzenemethanol ≥97%

Supplier: AOBChem USA

Warning

241854-1G 241854-500MG 241854-5G 241854-25G
77467-254EA 415.09 USD
77467-254 77479-978 77490-286 77500-462
α-Hexyl-2,4-dimethylbenzenemethanol ≥97%
α-Hexyl-2,4-dimethylbenzenemethanol

Precaución: For research use only

Formula: C₁₅H₂₄O
MW: 220.35 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19602393
CAS Number: 1339800-96-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR