6-(2-Chlorophenyl)-3-pyridinamine ≥95%

Supplier: AOBChem USA

Warning

239987-1G 239987-500MG 239987-5G 239987-25G
77467-234EA 300.76 USD
77467-234 77479-958 77490-264 77500-442
6-(2-Chlorophenyl)-3-pyridinamine ≥95%
6-(2-Chlorophenyl)-3-pyridinamine

Precaución: For research use only

Formula: C₁₁H₉ClN₂
Storage Temperature: Ambient
CAS Number: 1119089-76-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR