2-Fluoro-6-(methylthio)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

23946-1G 23946-500MG 23946-5G 23946-25G
77467-230EA 166.86 USD
77467-230 77479-954 77490-260 77500-438
2-Fluoro-6-(methylthio)benzaldehyde ≥97%
2-Fluoro-6-(methylthio)benzaldehyde

Precaución: For research use only

Formula: C₈H₇FOS
MW: 170.204 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22543676
CAS Number: 153798-06-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR