2-Dimethylamino-5-pyridinemethanol ≥97%

Supplier: AOBChem USA

Warning

23809-1G 23809-500MG 23809-5G 23809-25G
77467-226EA 111.24 USD
77467-226 77479-950 77490-256 77500-434
2-Dimethylamino-5-pyridinemethanol ≥97%
2-Dimethylamino-5-pyridinemethanol

Precaución: For research use only

Formula: C₈H₁₂N₂O
MW: 152.194 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08236841
CAS Number: 20173-74-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR