2-Methoxy-4-(methylthio)pyridine-5-boronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

23756-1G 23756-500MG 23756-5G
77467-222EA 455.26 USD
77467-222 77479-946 77490-252
2-Methoxy-4-(methylthio)pyridine-5-boronic acid pinacol ester ≥97%
2-Methoxy-4-(methylthio)pyridine-5-boronic acid pinacol ester

Precaución: For research use only

Formula: C₁₃H₂₀BNO₃S
Storage Temperature: Refrigerator
MDL Number: MFCD22543670
CAS Number: 1451392-19-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR