3,4-Dibromo-2,5-bis(methylthio)thiophene ≥95%

Supplier: AOBChem USA

Warning

23722-1G 23722-500MG 23722-5G 23722-25G
77467-218EA 339.9 USD
77467-218 77479-942 77490-248 77500-428
3,4-Dibromo-2,5-bis(methylthio)thiophene ≥95%
3,4-Dibromo-2,5-bis(methylthio)thiophene

Precaución: For research use only

Formula: C₆H₆Br₂S₃
MW: 334.115 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00602720
CAS Number: 24444-81-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR