2,5-Bis(methylthio)thiophene ≥95%

Supplier: AOBChem USA

Warning

23719-1G 23719-500MG 23719-5G 23719-25G
77467-214EA 415.09 USD
77467-214 77479-938 77490-244 77500-424
2,5-Bis(methylthio)thiophene ≥95%
2,5-Bis(methylthio)thiophene

Precaución: For research use only

Formula: C₆H₈S₃
MW: 176.323 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00550158
CAS Number: 50878-66-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR