3-Bromo-2-(methylthio)thiophene ≥97%

Supplier: AOBChem USA

Warning

23658-1G 23658-500MG 23658-5G 23658-25G
77467-210EA 82.4 USD
77467-210 77479-934 77490-240 77500-420
3-Bromo-2-(methylthio)thiophene ≥97%
3-Bromo-2-(methylthio)thiophene

Precaución: For research use only

Formula: C₅H₅BrS₂
Storage Temperature: Ambient
MDL Number: MFCD22056778
CAS Number: 66768-75-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR