3-Bromo-2-formylthiophene-5-boronic acid ≥97%

Supplier: AOBChem USA

Warning

23535-1G 23535-500MG 23535-5G 23535-25G
77467-202EA 195.7 USD
77467-202 77479-926 77490-232 77500-410
3-Bromo-2-formylthiophene-5-boronic acid ≥97%
3-Bromo-2-formylthiophene-5-boronic acid

Precaución: For research use only

Formula: C₅H₄BBrO₃S
Storage Temperature: Ambient
MDL Number: MFCD23713095
CAS Number: 1451393-26-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR