α-Ethenyl-2-fluoro-5-iodobenzenemethanol ≥97%

Supplier: AOBChem USA

Warning

235154-1G 235154-500MG 235154-5G 235154-25G
77467-198EA 397.58 USD
77467-198 77479-922 77490-228 77500-408
α-Ethenyl-2-fluoro-5-iodobenzenemethanol ≥97%
α-Ethenyl-2-fluoro-5-iodobenzenemethanol

Precaución: For research use only

Formula: C₉H₈FIO
Storage Temperature: Ambient
MDL Number: MFCD34870246
CAS Number: 2352791-73-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR