α-Ethenyl-2,4-difluorobenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

235140-1G 235140-500MG 235140-5G 235140-25G
77467-194EA 380.07 USD
77467-194 77479-918 77490-224 77500-406
α-Ethenyl-2,4-difluorobenzenemethanol ≥95%
α-Ethenyl-2,4-difluorobenzenemethanol

Precaución: For research use only

Formula: C₉H₈F₂O
Storage Temperature: Ambient
MDL Number: MFCD21232132
CAS Number: 149946-81-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR