α-Ethenyl[1,1'-biphenyl]-3-methanol ≥95%

Supplier: AOBChem USA

Warning

235096-1G 235096-500MG 235096-5G
77467-190EA 696.28 USD
77467-190 77479-914 77490-220
α-Ethenyl[1,1'-biphenyl]-3-methanol ≥95%
α-Ethenyl[1,1'-biphenyl]-3-methanol

Precaución: For research use only

Formula: C₁₅H₁₄O
MW: 210.27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34870233
CAS Number: 2353174-13-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR