α-Ethenyl-2,3-difluorobenzenemethanol ≥97%

Supplier: AOBChem USA

Warning

235081-1G 235081-500MG 235081-5G
77467-186EA 751.9 USD
77467-186 77479-910 77490-216
α-Ethenyl-2,3-difluorobenzenemethanol ≥97%
α-Ethenyl-2,3-difluorobenzenemethanol

Precaución: For research use only

Formula: C₉H₈F₂O
MW: 170.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21232155
CAS Number: 149946-80-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR