α-Ethenyl-2,6-difluorobenzenemethanol ≥97%

Supplier: AOBChem USA

Warning

235067-1G 235067-500MG 235067-5G
77467-182EA 751.9 USD
77467-182 77479-906 77490-212
α-Ethenyl-2,6-difluorobenzenemethanol ≥97%
α-Ethenyl-2,6-difluorobenzenemethanol

Precaución: For research use only

Formula: C₉H₈F₂O
MW: 170.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21232116
CAS Number: 149946-83-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR