α-Ethenyl-3-(1-methylethyl)benzenemethanol ≥95%

Supplier: AOBChem USA

Warning

234964-1G 234964-500MG 234964-5G
77467-178EA 553.11 USD
77467-178 77479-902 77490-206
α-Ethenyl-3-(1-methylethyl)benzenemethanol ≥95%
α-Ethenyl-3-(1-methylethyl)benzenemethanol

Precaución: For research use only

Formula: C₁₂H₁₆O
MW: 176.26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34870201
CAS Number: 2229153-34-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR