α-Ethenyl-2-ethoxybenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

234952-1G 234952-500MG 234952-5G 234952-25G
77467-174EA 380.07 USD
77467-174 77479-898 77490-202 77500-400
α-Ethenyl-2-ethoxybenzenemethanol ≥95%
α-Ethenyl-2-ethoxybenzenemethanol

Precaución: For research use only

Formula: C₁₁H₁₄O₂
Storage Temperature: Ambient
MDL Number: MFCD21232061
CAS Number: 1495685-78-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR