α-Ethenyl-3,5-dimethylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

234921-1G 234921-500MG 234921-5G 234921-25G
77467-170EA 380.07 USD
77467-170 77479-894 77490-198 77500-396
α-Ethenyl-3,5-dimethylbenzenemethanol ≥95%
α-Ethenyl-3,5-dimethylbenzenemethanol

Precaución: For research use only

Formula: C₁₁H₁₄O
Storage Temperature: Ambient
MDL Number: MFCD21232154
CAS Number: 1494121-30-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR