α-Ethenyl-3-fluorobenzenemethanol ≥97%

Supplier: AOBChem USA

Warning

234915-1G 234915-500MG 234915-5G
77467-166EA 751.9 USD
77467-166 77479-890 77490-194
α-Ethenyl-3-fluorobenzenemethanol ≥97%
α-Ethenyl-3-fluorobenzenemethanol

Precaución: For research use only

Formula: C₉H₉FO
Storage Temperature: Ambient
MDL Number: MFCD21232108
CAS Number: 38340-15-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR