α-Ethenyl-2-methylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

234903-1G 234903-500MG 234903-5G 234903-25G
77467-162EA 380.07 USD
77467-162 77479-886 77490-190 77500-392
α-Ethenyl-2-methylbenzenemethanol ≥95%
α-Ethenyl-2-methylbenzenemethanol

Precaución: For research use only

Formula: C₁₀H₁₂O
MW: 148.2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21232121
CAS Number: 39627-62-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR